6-Methyl-1H-pyrrolo[2,3-b]pyridine-5-carboxylic acid 95%

Supplier: Ambeed
Warning

A967393-1G A967393-250MG A967393-5G
77147-308EA 491.2 USD
77147-308 77147-310 77147-312
6-Methyl-1H-pyrrolo[2,3-b]pyridine-5-carboxylic acid 95%
6-Methyl-1H-pyrrolo[2,3-b]pyridine-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MW: 176.17 g/mol
MDL Number: MFCD09880165
CAS Number: 872355-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR