[1,2,4]Triazolo[1,5-a]pyrazine 95%

Supplier: Ambeed
Warning

A966591-1G A966591-250MG
77147-304EA 31.93 USD
77147-304 77147-306
[1,2,4]Triazolo[1,5-a]pyrazine 95%
[1,2,4]Triazolo[1,5-a]pyrazine

Precaución: for Research Use Only

Formula: C₅H₄N₄
MDL Number: MFCD09834988
CAS Number: 399-66-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR