(R)-(+)-2-[2-(Diphenylphosphino)phenyl]-4-(1-methylethyl)-4,5-dihydrooxazole ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (R)-iPr-PHOX, (R)-2-(2-(Diphenylphosphino)phenyl)-4-isopropyl-4,5-dihydrooxazole

A965930-100MG A965930-1G A965930-250MG
77147-302EA 31.83 USD
77147-302 77573-912 77573-914
(R)-(+)-2-[2-(Diphenylphosphino)phenyl]-4-(1-methylethyl)-4,5-dihydrooxazole ≥98%, ee 99%
(R)-(+)-2-[2-(Diphenylphosphino)phenyl]-4-(1-methylethyl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₄H₂₄NOP
MW: 373.4200 g/mol
MDL Number: MFCD02684553
CAS Number: 164858-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR