(S)-4-(Hydroxymethyl)oxazolidin-2-one 98%

Supplier: Ambeed
Danger

A963522-100MG A963522-1G A963522-250MG A963522-5G
77147-292EA 26.53 USD
77147-292 77147-294 77147-296 77573-906
(S)-4-(Hydroxymethyl)oxazolidin-2-one 98%
(S)-4-(Hydroxymethyl)oxazolidin-2-one

Precaución: for Research Use Only

Formula: C₄H₇NO₃
MW: 117.10 g/mol
MDL Number: MFCD11847630
CAS Number: 144542-44-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR