trans-2-(pyridin-4-yl)cyclopropanecarboxylic acid 97%

Supplier: Ambeed
Warning

A962576-1G A962576-250MG
77147-288EA 235.52 USD
77147-288 77147-290
trans-2-(pyridin-4-yl)cyclopropanecarboxylic acid 97%
trans-2-(pyridin-4-yl)cyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₉H₉NO₂
MW: 163.17 g/mol
MDL Number: MFCD24369465
CAS Number: 801149-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR