Tetrabutylphosphonium tetraphenylborate 98%

Supplier: Ambeed
Warning

A955253-25G A955253-5G
77147-284EA 82.4 USD
77147-284 77147-286
Tetrabutylphosphonium tetraphenylborate 98%
Tetrabutylphosphonium tetraphenylborate

Precaución: for Research Use Only

Formula: C₄₀H₅₆BP
MDL Number: MFCD04038112
CAS Number: 29089-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR