4-Chloro-3-nitrobenzamide 98%

Supplier: Ambeed
Warning

A953768-100G A953768-1G A953768-25G A953768-5G
77147-276EA 499.69 USD
77147-276 77147-278 77147-280 77147-282
4-Chloro-3-nitrobenzamide 98%
4-Chloro-3-nitrobenzamide

Precaución: for Research Use Only

Formula: C₇H₅ClN₂O₃
MDL Number: MFCD00017134
CAS Number: 16588-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR