N-Methylallylamine 98%

Supplier: Ambeed
Danger

Synonyms: N-Allylmethylamine

A947648-10G A947648-25G A947648-5G
77147-270EA 16.98 USD
77147-270 77147-272 77147-274
N-Methylallylamine 98%
N-Methylallylamine

Precaución: for Research Use Only

Formula: C₄H₉N
MW: 71.12 g/mol
MDL Number: MFCD00008641
CAS Number: 627-37-2
UN: 3286
ADR: 3,6.1,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR