2-[2-(sec-Butyl)phenoxy]acetic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-(2-(sec-Butyl)phenoxy)acetic acid

A946424-1G A946424-250MG A946424-5G
77147-264EA 27.59 USD
77147-264 77147-266 77147-268
2-[2-(sec-Butyl)phenoxy]acetic acid 95%
2-[2-(sec-Butyl)phenoxy]acetic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
MW: 208.26 g/mol
MDL Number: MFCD00268165
CAS Number: 76343-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR