4-Bromo-2-fluoro-3-methoxybenzaldehyde 98%

Supplier: Ambeed
Danger

A944628-1G A944628-250MG
77147-244EA 96.55 USD
77147-244 77147-246
4-Bromo-2-fluoro-3-methoxybenzaldehyde 98%
4-Bromo-2-fluoro-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
MW: 233.03 g/mol
MDL Number: MFCD27578163
CAS Number: 1820614-17-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR