(2S,4S)-(-)-2,4-Bis(diphenylphosphino)pentane 98%

Supplier: Ambeed
Warning

Synonyms: (2S,4S)-Pentane-2,4-diylbis(diphenylphosphine), (S,S)-BDPP

A943826-100MG A943826-250MG A943826-5G A943826-1G
77147-238EA 31.83 USD
77147-238 77147-240 77147-242 77618-606
(2S,4S)-(-)-2,4-Bis(diphenylphosphino)pentane 98%
(2S,4S)-(-)-2,4-Bis(diphenylphosphino)pentane

Precaución: for Research Use Only

Formula: C₂₉H₃₀P₂
MW: 440.50 g/mol
MDL Number: MFCD00058923
CAS Number: 77876-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR