Neopentyl glycol dimethacrylate 98% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A942536-1000G A942536-100G A942536-25G
77147-232EA 162.74 USD
77147-232 77147-234 77147-236
Neopentyl glycol dimethacrylate 98% stabilized
Neopentyl glycol dimethacrylate

Precaución: for Research Use Only

Formula: C₁₃H₂₀O₄
MDL Number: MFCD00048120
CAS Number: 1985-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR