(2S,5R)-N-Boc-5-methylpyrrolidine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A941647-100MG A941647-1G A941647-250MG
77147-220EA 187.78 USD
77147-220 77147-222 77147-224
(2S,5R)-N-Boc-5-methylpyrrolidine-2-carboxylic acid 95%
(2S,5R)-N-Boc-5-methylpyrrolidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MW: 229.27 g/mol
MDL Number: MFCD11111135
CAS Number: 160033-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR