3-(3,5-Difluorophenyl)oxetan-3-ol 97%

Supplier: Ambeed
Warning

A932975-100MG A932975-1G
77147-210EA 20.6 USD
77147-210 77147-212
3-(3,5-Difluorophenyl)oxetan-3-ol 97%
3-(3,5-Difluorophenyl)oxetan-3-ol

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
MDL Number: MFCD23703311
CAS Number: 1395281-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR