sulfino-L-alanine hydrate 98%

Supplier: Ambeed
Warning

Synonyms: L-Cysteinesulphinic acid monohydrate

A922755-1MG A922755-50MG
77147-198EA 21.22 USD
77147-198 77147-200
sulfino-L-alanine hydrate 98%
sulfino-L-alanine hydrate

Precaución: for Research Use Only

Formula: C₃H₉NO₅S
MW: 171.17 g/mol
MDL Number: MFCD00150732
CAS Number: 207121-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR