tert-Butyl methyl(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)carbamate 95%

Supplier: Ambeed
Warning

A922352-1G A922352-250MG A922352-5G
77147-192EA 201.58 USD
77147-192 77147-194 77147-196
tert-Butyl methyl(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)carbamate 95%
tert-Butyl methyl(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₇H₂₇BN₂O₄
MW: 334.22 g/mol
MDL Number: MFCD18382785
CAS Number: 1254381-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR