2-Chloro-5-methoxypyridine-4-boronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

A920531-1G A920531-250MG A920531-5G
77147-186EA 156.56 USD
77147-186 77147-188 77147-190
2-Chloro-5-methoxypyridine-4-boronic acid pinacol ester 97%
2-Chloro-5-methoxypyridine-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BClNO₃
MDL Number: MFCD18723303
CAS Number: 2096336-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR