2-Chloro-3-fluorotoluene 98%

Supplier: Ambeed
Warning

A918092-1G A918092-5G
77147-182EA 32.96 USD
77147-182 77147-184
2-Chloro-3-fluorotoluene 98%
2-Chloro-3-fluorotoluene

Precaución: for Research Use Only

Formula: C₇H₆ClF
MDL Number: MFCD08445246
CAS Number: 116850-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR