2,5,8,11,14-Pentaoxaheptadecan-17-oic acid 98%

Supplier: Ambeed
Warning

A915395-100G A915395-1G A915395-250MG A915395-25G A915395-5G
77147-170EA 1873.55 USD
77147-170 77147-172 77147-174 77147-176 77147-178
2,5,8,11,14-Pentaoxaheptadecan-17-oic acid 98%
2,5,8,11,14-Pentaoxaheptadecan-17-oic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₄O₇
MW: 280.31 g/mol
MDL Number: MFCD20926398
CAS Number: 81836-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR