4-Cyclopropyl-2-fluoro-6-methylbenzamide 97%

Supplier: Ambeed
Warning

A910651-1G A910651-250MG
77147-154EA 163.77 USD
77147-154 77147-156
4-Cyclopropyl-2-fluoro-6-methylbenzamide 97%
4-Cyclopropyl-2-fluoro-6-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₂FNO
MDL Number: MFCD20726776
CAS Number: 1242156-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR