1,5-Diamino-2,4-dichlorobenzene 95%

Supplier: Ambeed
Warning

A906123-100MG A906123-1G A906123-250MG
77147-144EA 67.99 USD
77147-144 77147-146 77147-148
1,5-Diamino-2,4-dichlorobenzene 95%
1,5-Diamino-2,4-dichlorobenzene

Precaución: for Research Use Only

Formula: C₆H₆Cl₂N₂
MW: 177.03 g/mol
MDL Number: MFCD00456308
CAS Number: 20248-64-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR