1-(2-Fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)-4-methylpiperazine dihydrochloride 97%

Supplier: Ambeed
Warning

A901519-1G A901519-250MG A901519-5G
77147-120EA 143.17 USD
77147-120 77147-122 77147-124
1-(2-Fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)-4-methylpiperazine dihydrochloride 97%
1-(2-Fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)-4-methylpiperazine dihydrochloride

Precaución: for Research Use Only

Formula: C₁₈H₃₀BCl₂FN₂O₂
MDL Number: MFCD20529512
CAS Number: 2096334-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR