2-(3,5-Difluorophenyl)ethanol 98%
Supplier: Ambeed
|
Warning
|
A899971-1G
A899971-25G
A899971-5G
77147-116EA
19.57
USD
77147-116
77147-118
77573-808
2-(3,5-Difluorophenyl)ethanol 98%
2-(3,5-Difluorophenyl)ethanol
Precaución: for Research Use Only
Formula:
C₈H₈F₂O |
MDL Number:
MFCD06201148 CAS Number: 467223-90-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...