Bifonazole 97%

Supplier: Ambeed
Warning

Synonyms: 1-(alpha,4-Diphenylbenzyl)imidazole, 1-([1,1'-Biphenyl]-4-yl(phenyl)methyl)-1H-imidazole

A899029-100G A899029-1G A899029-250MG A899029-25G A899029-5G
77147-102EA 62.83 USD
77147-102 77147-104 77147-106 77147-108 77147-110
Bifonazole 97%
Bifonazole

Precaución: for Research Use Only

Formula: C₂₂H₁₈N₂
MDL Number: MFCD00865567
CAS Number: 60628-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR