2-Chloro-4-methyl-5-nitrobenzoic acid 97%
Supplier: Ambeed
|
Warning
|
A896885-1G
A896885-250MG
A896885-5G
77147-096EA
61.8
USD
77147-096
77147-098
77595-438
2-Chloro-4-methyl-5-nitrobenzoic acid 97%
2-Chloro-4-methyl-5-nitrobenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₆ClNO₄ |
MDL Number:
MFCD27940870 CAS Number: 101580-96-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...