Diammonium ethylenediaminetetraacetate monohydrate 98%

Supplier: Ambeed
Warning

Synonyms: Diammonium EDTA, Ammonium 2,2'-((2-(bis(carboxymethyl)amino)ethyl)azanediyl)diacetate hydrate, Ethylenediaminetetraacetic Acid Diammonium Salt

A894346-25G A894346-5G
77147-084EA 22.66 USD
77147-084 77147-086
Diammonium ethylenediaminetetraacetate monohydrate 98%
Diammonium ethylenediaminetetraacetate monohydrate

Precaución: for Research Use Only

Formula: C₁₀H₂₄N₄O₉
MDL Number: MFCD00150463
CAS Number: 304675-80-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR