Ethyl 2,4-dibromo-1H-imidazole-5-carboxylate 97%

Supplier: Ambeed
Danger

A892880-100MG A892880-1G A892880-250MG A892880-5G
77147-076EA 19.1 USD
77147-076 77147-078 77147-080 77147-082
Ethyl 2,4-dibromo-1H-imidazole-5-carboxylate 97%
Ethyl 2,4-dibromo-1H-imidazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆Br₂N₂O₂
MW: 297.93 g/mol
MDL Number: MFCD29059195
CAS Number: 74478-96-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR