Semagacestat 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Hydroxy-3-methyl-N-((S)-1-(((S)-3-methyl-2-oxo-2,3,4,5-tetrahydro-1H-benzo[d]azepin-1-yl)amino)-1-oxopropan-2-yl)butanamide

A892738-10MG A892738-1MG A892738-50MG A892738-5MG
77147-068EA 57.29 USD
77147-068 77147-070 77147-072 77147-074
Semagacestat 98%
Semagacestat

Precaución: for Research Use Only

Formula: C₁₉H₂₇N₃O₄
MW: 361.44 g/mol
MDL Number: MFCD25542369
CAS Number: 425386-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR