(4S)-3-[(Benzyloxy)carbonyl]-2-oxoimidazolidine-4-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: (S)-3-((Benzyloxy)carbonyl)-2-oxoimidazolidine-4-carboxylic acid

A891873-1G A891873-250MG A891873-25G A891873-5G
77147-054EA 45.62 USD
77147-054 77147-056 77147-058 77147-060
(4S)-3-[(Benzyloxy)carbonyl]-2-oxoimidazolidine-4-carboxylic acid 97%
(4S)-3-[(Benzyloxy)carbonyl]-2-oxoimidazolidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O₅
MW: 264.24 g/mol
MDL Number: MFCD00192373
CAS Number: 59760-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR