m-PEG7-amine 98%

Supplier: Ambeed
Warning

A890713-100MG A890713-250MG A890713-1G
77147-050EA 56.65 USD
77147-050 77147-052 77686-930
m-PEG7-amine 98%
m-PEG7-amine

Precaución: for Research Use Only

Formula: C₁₅H₃₃NO₇
MDL Number: MFCD13184965
CAS Number: 170572-38-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR