4-Chloro-1-methyl-1H-pyrazole-3-carboxylic acid 98%

Supplier: Ambeed
Danger

A888815-250MG A888815-5G
77147-044EA 36.08 USD
77147-044 77147-046
4-Chloro-1-methyl-1H-pyrazole-3-carboxylic acid 98%
4-Chloro-1-methyl-1H-pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O₂
MDL Number: MFCD01821187
CAS Number: 84547-85-3
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR