Methyl 2-hydroxy-2-(1-methyl-1H-pyrazol-4-yl)acetate 97%

Supplier: Ambeed
Warning

A886911-100MG A886911-1G A886911-250MG
77147-034EA 79.31 USD
77147-034 77147-036 77147-038
Methyl 2-hydroxy-2-(1-methyl-1H-pyrazol-4-yl)acetate 97%
Methyl 2-hydroxy-2-(1-methyl-1H-pyrazol-4-yl)acetate

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₃
MDL Number: MFCD16693842
CAS Number: 1251328-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR