(R)-1-(Naphthalen-1-yl)ethanol 95%

Supplier: Ambeed
Warning

A885090-100MG A885090-1G A885090-250MG
77147-026EA 50.46 USD
77147-026 77147-028 77147-030
(R)-1-(Naphthalen-1-yl)ethanol 95%
(R)-1-(Naphthalen-1-yl)ethanol

Precaución: for Research Use Only

Formula: C₁₂H₁₂O
MDL Number: MFCD00077830
CAS Number: 42177-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR