Methyl 6-chloro-3-methylbenzo[b]thiophene-2-carboxylate 97%

Supplier: Ambeed
Warning

A883924-100MG A883924-1G
77147-016EA 42.44 USD
77147-016 77147-018
Methyl 6-chloro-3-methylbenzo[b]thiophene-2-carboxylate 97%
Methyl 6-chloro-3-methylbenzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₉ClO₂S
MW: 240.70 g/mol
MDL Number: MFCD26096488
CAS Number: 1415968-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR