6-Bromo-8-chloro-3-methyl-[1,2,4]triazolo[4,3-a]pyrazine 97%

Supplier: Ambeed
Warning

A879842-100MG A879842-1G A879842-250MG A879842-5G
77147-006EA 56.64 USD
77147-006 77147-008 77147-010 77147-012
6-Bromo-8-chloro-3-methyl-[1,2,4]triazolo[4,3-a]pyrazine 97%
6-Bromo-8-chloro-3-methyl-[1,2,4]triazolo[4,3-a]pyrazine

Precaución: for Research Use Only

Formula: C₆H₄BrClN₄
MDL Number: MFCD28405548
CAS Number: 1610045-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR