Methyl 1-(5-bromopyrimidin-2-yl)cyclopropanecarboxylate 97%

Supplier: Ambeed
Warning

A878617-100MG A878617-1G A878617-250MG
77147-000EA 142.17 USD
77147-000 77147-002 77147-004
Methyl 1-(5-bromopyrimidin-2-yl)cyclopropanecarboxylate 97%
Methyl 1-(5-bromopyrimidin-2-yl)cyclopropanecarboxylate

Precaución: for Research Use Only

Formula: C₉H₉BrN₂O₂
MW: 257.08 g/mol
MDL Number: MFCD25542298
CAS Number: 1447607-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR