10H-Phenothiazine 5-oxide 95%

Supplier: Ambeed
Warning

A871333-1G A871333-5G
77146-984EA 120.51 USD
77146-984 77146-986
10H-Phenothiazine 5-oxide 95%
10H-Phenothiazine 5-oxide

Precaución: for Research Use Only

Formula: C₁₂H₉NOS
MDL Number: MFCD00053824
CAS Number: 1207-71-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR