2,4-Difluorobenzhydrazide 98%

Supplier: Ambeed
Warning

A866999-1G A866999-5G
77146-974EA 15.45 USD
77146-974 77146-976
2,4-Difluorobenzhydrazide 98%
2,4-Difluorobenzhydrazide

Precaución: for Research Use Only

Formula: C₇H₆F₂N₂O
MDL Number: MFCD00173685
CAS Number: 118737-62-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR