4-(1-Amino-1-carboxyethyl)benzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: RS-MCPG

A863113-10MG A863113-1MG A863113-25MG A863113-50MG A863113-5MG
77146-964EA 74.16 USD
77146-964 77146-966 77146-968 77146-970 77146-972
4-(1-Amino-1-carboxyethyl)benzoic acid 98%
4-(1-Amino-1-carboxyethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₄
MW: 209.2 g/mol
MDL Number: MFCD00210205
CAS Number: 146669-29-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR