4-Fluoro-2-methylbenzenesulfonyl chloride 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
4-Fluoro-2-methylbenzene-1-sulphonyl chloride
A855976-1G
A855976-250MG
A855976-5G
77146-938EA
27.59
USD
77146-938
77146-940
77146-942
4-Fluoro-2-methylbenzenesulfonyl chloride 95%
4-Fluoro-2-methylbenzenesulphonyl chloride
Precaución: for Research Use Only
Formula:
C₇H₆ClFO₂S MW: 208.64 g/mol |
MDL Number:
MFCD03094390 CAS Number: 7079-48-3 UN: 3261 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...