Formaldehyde sodium bisulfite 97%

Supplier: Ambeed
Warning

Synonyms: Sodium Hydroxymethanesulphonate, Hydroxymethanesulphonic acid sodium salt

A852769-100G A852769-25G
77146-934EA 10.61 USD
77146-934 77146-936
Formaldehyde sodium bisulfite 97%
Formaldehyde sodium bisulfite

Precaución: for Research Use Only

Formula: CH₄O₄S
MW: 134.08 g/mol
MDL Number: MFCD00044664
CAS Number: 870-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR