4-Bromo-1-phenylpyridin-2(1H)-one 97%

Supplier: Ambeed
Warning

A846723-100MG A846723-1G A846723-250MG
77146-898EA 45.62 USD
77146-898 77146-900 77146-902
4-Bromo-1-phenylpyridin-2(1H)-one 97%
4-Bromo-1-phenylpyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₁H₈BrNO
MW: 250.09 g/mol
MDL Number: MFCD29038523
CAS Number: 1935260-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR