(S)-tert-Butyl 2-(hydroxymethyl)-2,5-dihydro-1H-pyrrole-1-carboxylate 97%

Supplier: Ambeed
Warning

A846147-100MG A846147-1G
77146-884EA 57.29 USD
77146-884 77573-688
(S)-tert-Butyl 2-(hydroxymethyl)-2,5-dihydro-1H-pyrrole-1-carboxylate 97%
(S)-tert-Butyl 2-(hydroxymethyl)-2,5-dihydro-1H-pyrrole-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₃
MW: 199.24 g/mol
MDL Number: MFCD18074315
CAS Number: 205444-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR