(S)-(-)-1-Phenyl-1-propanol 98%

Supplier: Ambeed
Warning

Synonyms: (S)-1-phenyl-1-propanol, (S)-(-)-α-Ethylbenzyl alcohol

A841975-100MG A841975-250MG
77146-878EA 26.53 USD
77146-878 77146-880
(S)-(-)-1-Phenyl-1-propanol 98%
(S)-(-)-1-Phenyl-1-propanol

Precaución: for Research Use Only

Formula: C₉H₁₂O
MW: 136.19 g/mol
MDL Number: MFCD00066207
CAS Number: 613-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR