Ethyl-5-chlorothiophene-2-carboxylate 97%

Supplier: Ambeed
Warning

A841512-100G A841512-10G A841512-1G A841512-5G
77146-870EA 144.2 USD
77146-870 77146-872 77146-874 77146-876
Ethyl-5-chlorothiophene-2-carboxylate 97%
Ethyl 5-chlorothiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₇ClO₂S
MDL Number: MFCD00051732
CAS Number: 5751-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR