3-Amino-3-(3'-pyridyl)propionic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-3-(3-pyridyl)propionic Acid, (RS)-3-Amino-3-(3-pyridyl)-propionic acid

A840953-1G A840953-25G
77146-864EA 16.98 USD
77146-864 77146-866
3-Amino-3-(3'-pyridyl)propionic acid 95%
3-Amino-3-(3'-pyridyl)propionic acid

Precaución: for Research Use Only

Formula: C8H10N2O2
MW: 166.1800 g/mol
MDL Number: MFCD01549598
CAS Number: 62247-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR