(±)-1,1'-Bi-2-naphthol 98%

Supplier: Ambeed
Danger

A840497-10G A840497-5G A840497-25G A840497-100G
77146-860EA 6.18 USD
77146-860 77146-862 77552-568 77686-870
(±)-1,1'-Bi-2-naphthol 98%
(±)-1,1'-Bi-2-naphthol

Precaución: for Research Use Only

Formula: C₂₀H₁₄O₂
MDL Number: MFCD00004068
CAS Number: 602-09-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR