5-Bromo-4-hydroxynicotinonitrile 95%

Supplier: Ambeed
Warning

A839149-100MG A839149-1G
77146-848EA 93.36 USD
77146-848 77146-850
5-Bromo-4-hydroxynicotinonitrile 95%
5-Bromo-4-hydroxynicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₃BrN₂O
MW: 199.00 g/mol
MDL Number: MFCD28735815
CAS Number: 1160923-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR