5-Chloro-2,4-dinitrotoluene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Chloro-5-methyl-2,4-dinitrobenzene
A839045-1G
A839045-250MG
A839045-25G
A839045-5G
77146-840EA
22.28
USD
77146-840
77146-842
77146-844
77146-846
5-Chloro-2,4-dinitrotoluene 98%
5-Chloro-2,4-dinitrotoluene
Precaución: for Research Use Only
Formula:
C₇H₅ClN₂O₄ MW: 216.58 g/mol |
MDL Number:
MFCD00041235 CAS Number: 51676-74-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...