Indium(I) iodide 98%

Supplier: Ambeed
Danger

A836799-250MG A836799-1G
77146-828EA 9.27 USD
77146-828 77569-556
Indium(I) iodide 98%
Indium(I) iodide

Precaución: For research use only.

Formula: InI
MW: 241.72 g/mol
Boiling Pt: 711
Melting Pt: 351
MDL Number: MFCD00135543
CAS Number: 13966-94-4
UN: 3288
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR